missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid, ACS Grade, 95.0 to 98.0% (w/w), LabChem™

Supplier:  LabChem LC255503

 View more versions of this product

Catalog No. LC255503


Explore available promotions

Description

Description

Specifications

Specifications

Sulfuric Acid
10°C
288°C
100
HDPE Bottle
1840kg/m3
2.5 L
Liquid
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08
ACS
Colorless
7664-93-9
1840kg/m3
Passes Test
≤2ppm
Sulfur compounds
H2O4S
H2SO4
1830
Soluble in water
OS(O)(=O)=O
98.07
CHEBI:26836
95.0 to 98.0% (w/w)

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/docto

Danger

DOTInformation : Class 8: Corrosive

EINECSNumber : 231-639-5

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions