missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, Certified, 20.0% (v/v) ±0.5%, LabChem™

$83.50 - $170.40

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC256072
View Documents
LabChem
LC256072
1 L
Each for $83.50
Only null left
 
LC256074
View Documents
LabChem
LC256074
4 L
Each for $170.40
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O

Specifications

Colorless
1.21g/mL
Passes Test
1.21g/mL
Sulfur compounds; Thermal decomposition generates: Corrosive vapors
H2O4S
H2SO4
UN2796
Soluble in water
OS(O)(=O)=O
98.07
CHEBI:26836
20.0% ±0.5%(v/v)
Sulfuric Acid
7664-93-9,7732-18-5
70.85,29.15
Poly Bottle
Traceable to NIST
1 L
Liquid
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08
Certified
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'