missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfuric Acid, Certified, 50% (v/v) ±1%, LabChem™
Supplier: LabChem LC256402
Specifications
Sulfuric Acid, (1+1) | |
7664-93-9,7732-18-5 | |
59.23,40.77 | |
Amber Glass | |
Traceable to NIST | |
1 L | |
Liquid | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08 | |
Certified |
Colorless | |
1.49g/mL | |
Passes Test | |
1.49g/mL | |
Sulfur compounds; Thermal decomposition generates: Corrosive vapors | |
H2O4S | |
H2SO4 | |
UN1830 | |
Soluble in water | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
50% ±1% (v/v) |
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/docto
Danger
Recommended Storage : Room Temperature