missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfuric Acid Concentrate (Certified) ACS, Fisher Chemical
Each bottle contains exact amount of 10N concentrate required to make 1 liter of 1N or 10 liters of 0.1N (N/10) solution
Supplier: Thermo Fisher Scientific Formerly Fairlawn SA213
Specifications
0.03 | |
11°C | |
105°C | |
1.1g/mL | |
Glass Bottle | |
100 mL | |
Liquid | |
2796 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
Certified |
Sulfuric Acid Concentrate | |
7664-93-9,7732-18-5 | |
0.995 to 1.005N | |
Pass Test | |
<0.00120 mmHg | |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 |
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Safety and Handling
DOTInformation : Class 8: Corrosive