Learn More
Sulfuric acid, extra pure, fuming, 20-30% free SO3, Thermo Scientific Chemicals
$206.01 - $1219.77
Chemical Identifiers
CAS | 8014-95-7 |
---|---|
Molecular Formula | H2O7S2 |
Molecular Weight (g/mol) | 178.13 |
MDL Number | MFCD00137792 |
InChI Key | HIFJUMGIHIZEPX-UHFFFAOYSA-N |
SMILES | O=S(=O)=O.OS(O)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC419970250
|
Thermo Scientific Chemicals
419970250 |
25 g | Glass bottle |
Each for $206.01
|
|
||||
AC419975000
|
Thermo Scientific Chemicals
419975000 |
500 g | Glass bottle |
Each for $343.06
|
|
||||
AC419970025
|
Thermo Scientific Chemicals
419970025 |
2.5 kg | Glass bottle |
Each for $1,219.77
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
8014-95-7 | |
178.13 | |
HIFJUMGIHIZEPX-UHFFFAOYSA-N |
H2O7S2 | |
MFCD00137792 | |
O=S(=O)=O.OS(O)(=O)=O |
Specifications
Brown to Colorless | |
8014-95-7 | |
1.9200g/mL | |
0.01% max. | |
25 g | |
Liquid | |
MFCD00137792 | |
oleum, fuming sulfuric acid, sulfuric acid, fuming, oleum iodisum, sulfuric acid fuming, oleum fuming sulfuric acid, sulfuric acid, mixt with sulfur trioxide, sulfur trioxide, mixt. with sulfuric acid, sulfuric acid mixture with sulfur trioxide | |
HIFJUMGIHIZEPX-UHFFFAOYSA-N | |
sulfuric acid;sulfur trioxide | |
24681 | |
20 to 30% (free SO3) | |
Sulfuric acid |
2.0°C | |
138.0°C | |
Glass bottle | |
1.92 | |
H2O7S2 | |
H2SO4·SO3 | |
15, 9104 | |
Solubility in water: miscible. Other solubilities: soluble in ethanol | |
O=S(=O)=O.OS(O)(=O)=O | |
178.13 | |
178.14 | |
Extra Pure |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RTECSNumber : WS5605000
RUO – Research Use Only