missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 7664-93-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
PubChem CID | 1118 |
ChEBI | CHEBI:26836 |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A468-250
|
Seastar Chemicals Inc
S020301TFIF01 |
250 mL | FEP Bottle |
Each for $2,711.50
|
|
||||
A468-500
|
Seastar Chemicals Inc
S020301TFIF03 |
500 mL | FEP Bottle |
Each for $3,969.00
|
|
||||
A468-1
|
Seastar Chemicals Inc
S020301TFIF05 |
1 L | FEP Bottle |
Each for $5,203.92
|
|
||||
A4682
|
Seastar Chemicals Inc
S020301TFIF07 |
2 L | FEP Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Certificate of Lot Analysis included with each bottle.
- Ideal for analysis by ICP-MS and high-resolution ICP-MS.
- Evaluated for 65 metals at low-level ppt concentrations.
- Manufactured, purified, and packaged in a Class 1000 clean room.
- Packaged in specially cleaned Teflon FEP for PFA bottles to maintainproduct integrity and to eliminate metal leaching from glass bottles.
- Three year shelf life from time of manufacture available on COA.
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Specifications
0.3 | |
10°C | |
290°C | |
FEP Bottle | |
Certificate of Lot Analysis included with each bottle. | |
H2O4S | |
MFCD00064589 | |
15, 9104 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.07 | |
Sulfuric Acid |
Colorless to Brown | |
7664-93-9 | |
1.84g/cm³ | |
<0.001 mmHg at 20°C | |
250 mL | |
Liquid | |
1830 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
OPTIMA™ |
Safety and Handling
DOTInformation : DOT Class 8, : Corrosive