missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid Solution (0.1N (N/10)/Certified), Fisher Chemical™

Standardized to pH 4.4 at 25°C against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

Supplier:  Thermo Fisher Scientific FLSA2201

 View more versions of this product

Catalog No. SA220-1



Explore available promotions

Description

Description

Specifications

Specifications

1.0
0.1N (N/10)
0.0995 to 0.1005N
Poly Bottle
340°C
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
0.1N (N/10)
Sulfuric Acid Solution
7664-93-9,7732-18-5
Pass Test
LOT 84L Potassium acid phthalate
1 L
Liquid
2796
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
Certified

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Causes severe eye and skin irritation with possible burns. May be harmful if inhaled. Corrosive to metals. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:1

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Promotions

Promotions