missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuryl chloride, 1M soln. in dichloromethane, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals H31899AP

Catalog No. AAH31899AP


Explore available promotions

Description

Description

Sulfuryl chloride is often used as a source of chlorine. is widely used as a reagent in the conversion of C-H ? C-Cl adjacent to activating substituent such as carbonyls and sulfoxides. It also chlorinates alkanes, alkenes, alkynes, aromatics, ethers (such as tetrahydrofuran) and epoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfuryl chloride is often used as a source of chlorine. is widely used as a reagent in the conversion of C-H → C-Cl adjacent to activating substituent such as carbonyls and sulfoxides. It also chlorinates alkanes, alkenes, alkynes,
Specifications

Specifications

Sulfuryl chloride
500 mL
SO2Cl2
UN2922
Reacts with water.
ClS(Cl)(=O)=O
134.96
CHEBI:29291
1M soln. in dichloromethane
1.352
7791-25-5
Cl2O2S
MFCD00011451
sulfuryl chloride, sulfonyl chloride, sulphuryl dichloride, sulphuryl chloride, sulfonyl dichloride, sulfuric dichloride, sulfuric oxychloride, sulfurylchlorid, caswell no. 816, sulfurylchloride
YBBRCQOCSYXUOC-UHFFFAOYSA-N
sulfuryl dichloride
24648
134.96
Moisture sensitive

Chemical Identifiers

7791-25-5
134.96
YBBRCQOCSYXUOC-UHFFFAOYSA-N
24648
sulfuryl dichloride
Cl2O2S
MFCD00011451
sulfuryl chloride, sulfonyl chloride, sulphuryl dichloride, sulphuryl chloride, sulfonyl dichloride, sulfuric dichloride, sulfuric oxychloride, sulfurylchlorid, caswell no. 816, sulfurylchloride
CHEBI:29291
ClS(Cl)(=O)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H351
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.

P201-P202-P260-P264b-P271-P281-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H330-H335-H336-H350

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 232-245-6

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only