Learn More
Supel™-Inert Multi-Layer Foil Gas Sampling Bags, Sold by MilliporeSigma™ Supelco™
Ideal for sampling low molecular weight compounds. Two outer layers of aluminum film provide a great barrier to gases permeating through the walls of the bag.
Supplier:  Merck Emd Millipore 30279U
 View more versions of this product
Catalog No. 111010610
Only null left
Explore available promotions
Description
Description
  • Proven low-bleed Thermogreen™ LB-2 Septa installed in the valve fitting
  • Chemically inert with moisture and light protection
  • Not recommended for low ppm VOCs due to background levels
Specifications
Specifications
Chemically Inert with Light/Moisture Protection
Screw Cap Valve (SCV)
0.6 L
10
Supel
6 in.
Multi-layer Foil Gas Sampling Bags
NIOSH 6603, OSHA ID-172 (ID-210)
Sampling Low Molecular Weight Compounds Such as Carbon Monoxide (CO), Carbon Dioxide (CO2) and Permanent Gases. Also Suitable for Sampling Sulfur Compounds Such as Hydrogen Sulfide (H2S), Carbon Disulfide (CS2) and Mercaptans.
Multi-layer foil composed of two outer layers of aluminum film
Thermogreen™ LB-2 Septa
Chemicals
6 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC