MilliporeSigma™ Supelco™ Supelclean™ ENVI-Carb™/NH2 SPE Tube, Bed Wt. 1000 mg ENVI-Carb™/500 mg LC-NH2
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
$638.50 - $650.91
Specifications
Column Phase ENVI-Carb/LC-NH2
Column Type SPE
Bed Weight 1000 mg ENVI-Carb™/500 mg LC-NH2
For Use With (Application) Food and Beverages
Product Line Supelclean ENVI
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Quantity Price Quantity  
111005394
View Documents
MilliporeSigma™ Supelco™
54117U
6 mL 30/Pk.
Each for $650.91
111005393
View Documents
MilliporeSigma™ Supelco™
54118U
12 mL 20/Pk.
Each for $638.50
Description
Description
Retention Mechanism: Reversed-phase and anion-exchange. Sample Matrix Compatibility: Organic or aqueous solutions. Dual layer SPE tube that contains both Supelclean ENVI-Carb (upper layer) and LC-NH2 (lower layer) SPE sorbents (separated by PE frit). Developed to offer superior clean up when conducting multi-residue pesticide analysis from food (e.g. agricultural products, meats, etc.). . ENVI-carb has a strong affinity towards planar molecules, and can isolate/remove pigments (e.g., chlorophyll and carotinoids) and sterols commonly present in foods and natural products. Supelclean LC-NH2 is an aminopropyl phase that retains fatty acids, organic acids, and some polar pigments and sugars common in food matrices.
Specifications
Specifications
ENVI-Carb/LC-NH2
1000 mg ENVI-Carb™/500 mg LC-NH2
Supelclean ENVI
Polypropylene
SPE
Food and Beverages
Solid Phase Extraction Column
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Supelclean and ENVI are trademarks of Sigma-Aldrich Co. LLC