missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Metro™ Super Erecta 4-Shelf Industrial Wire Shelving Starter Unit, Metroseal Gray Epoxy
Super Erecta 4-Shelf Industrial Wire Shelving Starter Unit consist of four posts and four Super Erecta shelves.
Supplier: Metro™ SE243074K4S4
Description
- Warm gray look that blends with most environments.
- Superior rust protection than chrome wire shelving.
- Built-in Microban antimicrobial product protection.
- 12-year corrosion resistance warranty.
Specifications
29.875 in. | |
Starter Unit | |
189.87 cm | |
24 x 30 in. | |
22.3 kg | |
907 kg | |
4 | |
Not Autoclavable | |
2000 lb. | |
Epoxy-coated Steel |
Product will be free from defects in material and workmanship under normal use, service and maintenance for a period of one (1) year from the date of shipment of the Product by Seller, unless otherwise specified by Seller in writing. Metroseal shelves and posts also carry a twelve (12) year warranty against rust formation and corrosion. | |
All Laboratory and Clean Room Non-Autoclavable Applications | |
74 in. | |
24.125 in. | |
49.3 lb. | |
612.775 mm | |
Super Erecta Shelving | |
758.825 mm | |
Gray |