missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Metro™ Super Erecta 4-Shelf Industrial Wire Shelving Starter Unit, Metroseal Gray Epoxy

Super Erecta 4-Shelf Industrial Wire Shelving Starter Unit consist of four posts and four Super Erecta shelves.

Supplier:  Metro™ SE213674K4S4

 View more versions of this product

Catalog No. 13460436


Explore available promotions

Description

Description

  • Warm gray look that blends with most environments.
  • Superior rust protection than chrome wire shelving.
  • Built-in Microban antimicrobial product protection.
  • 12-year corrosion resistance warranty.
Specifications

Specifications

35.875 in.
Starter Unit
189.87 cm
21 x 36 in.
26.6 kg
907 kg
4
Not Autoclavable
2000 lb.
Epoxy-coated Steel
Product will be free from defects in material and workmanship under normal use, service and maintenance for a period of one (1) year from the date of shipment of the Product by Seller, unless otherwise specified by Seller in writing. Metroseal shelves and posts also carry a twelve (12) year warranty against rust formation and corrosion.
All Laboratory and Clean Room Non-Autoclavable Applications
74 in.
21.125 in.
58.7 lb.
536.575 mm
Super Erecta Shelving
911.225 mm
Gray
Videos
SDS
Documents

Documents

Promotions

Promotions