missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ SuperChip™ Microarray Slides, Epoxysilane
Choose from several standard surface chemistries and custom coatings with Epredia™ SuperChip™ Microarray Slides.
$443.18 - $993.63
Specifications
Description | SuperChip Microarray Slides |
---|---|
Color | Plain |
Material | Fluorescence Glass |
Type | Superchip Microarray Slide |
Length (Metric) | 70 mm |
Catalog Number | Mfr. No. | Coating | Enhanced Surface | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Coating | Enhanced Surface | Price | Quantity | ||||
22-037-210
|
Epredia™
C185131M20 |
Aminopropylsilane | No |
N/A
|
|
||||
22-037-216
|
Epredia™
C405257M20 |
Poly-L-Lysine | No |
N/A
|
|
||||
Description
Aminopropylsilane-Coated Slides:- Ideal for microarrays fabricated with PCR products or proteins as probes
Poly-L-Lysine-Coated Slides:
- Offer an economical alternative to aminopropylsilane
Aldehyde-Silane-Coated Slides:
- Provide a uniform coating of aldehydes for the attachment of DNA and protein molecules
Epoxysilane-Coated Slides:
- Utilize a coating of epoxide groups for the attachment of amine-modified DNA, proteins, peptides, etc. with an amine end
SuperChip Advance:
- Nitrocellulose-coated slides for use in reverse phase arrays or protein microarray
SmartPlex:
- Multiplexing microarray platform SmartPlex allows for easy printing of arrays onto glass prior to attaching 96-well upper structure for multiplexing
Product Details:
- Consistent surface coating to provide a uniform DNA or protein immobilizing surface
- Optimal amount of probe binded while minimizing nonspecific binding of labeled targets
- Many available with an enhanced surface (ES) treatment that produces a more uniform, slightly larger spot size without changing surface chemistry
- Compatible with standard microarray spotting and detection systems
- Ready for immediate use or may be stored at room temperature for up to a year (six months for Poly-L-Lysine)
- Printable area: 20 × 70mm
- Secure no-contact mBox™ packaging, vacuum sealed in a foil barrier bag
Specifications
SuperChip Microarray Slides | |
Fluorescence Glass | |
70 mm | |
20 mm |
Plain | |
Superchip Microarray Slide | |
70 x 20 mm | |
No |