missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva Superdex™ 200 Increase 5/150 GL Prepacked Tricorn™ Columns

For size exclusion chromatography and analysis of proteins with molecular weights between 10,000 and 600,000. Cytiva Lifescience™ Superdex™ 200 Increase 5/150 GL Prepacked Tricorn™ Columns are designed for small scale preparative purification.

Supplier:  Cytiva 28990945

Catalog No. 45-002-571


Explore available promotions

Description

Description

  • For rapid purity checks and different experimental conditions screenings
  • Prepacked with Superdex 200 Increase media
  • Composite of cross-linked high-flow agarose and dextran base matrix for high flow rates
  • Smaller bead size of average 8.6μm for increased resolution
  • Flow Rate: <0.75mL/min1
  • Bed Volume: Approximately 3mL
  • Column 1.D.: 5mm
  • Columns are supplied with two fingertight connectors 1/16 in. male for connection to AKTA™ or other system

Specifications

Specifications

4 to 50μL
Superdex 200 Increase
Rapid purity checks and quick screenings
Cross-linked agarose and dextran
Videos
SDS
Documents

Documents

Promotions

Promotions