Cytiva Superdex 75 Increase Prepacked Column, 3.2/300
Use this column for rapid purity check and screening of proteins with molecular weights between Mr 3,000 and 70,000, such as recombinant tagged proteins. Cytiva Superdex 75 Increase Prepacked Columns are ideal when time is limited and low buffer and sample consumption is important.
Supplier: Cytiva 29148723
Description
- Up to 50% higher resolution compared with predecessor Superdex 75, for improved purity and analysis results
- Runtime reduced down to one third compared with predecessor Superdex 75, with maintained resolution for results faster
- Versatile use in both preparative and analytical applications, especially with recombinant tagged proteins
- Tolerance to repeated harsh cleaning procedures at high pH ensures long column lifetime and minimizes carryover
- Higher lot-to-lot consistency compared with predecessor Superdex 75
Specifications
High resolution size exclusion chromatography (SEC) in micro scale (4 to 50μL sample volume) | |
∼2.4 mL | |
4°C to 30°C |