missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ SuperFastPrep-2™ Portable Bead Beating Grinder and Lysis System, 2 x 2 mL, Battery Operated
SuperFastPrep-2™ Portable Bead Beating Grinder and Lysis System is used for Sample Preparation, lysis, homogenization and grinding of virtually any type of biological sample. It is handheld and compatible with most 2 mL tubes containing lysis matrix.
Supplier: MP Biomedicals™ 116012500
Description
- Product Overview: The SuperFastPrep2 is a compact, cordless, hand-held sample disruption instrument designed to quickly and efficiently homogenize, grind, or lyse biological and environmental samples. It can be used at remote locations and in most weather conditions for the immediate collection, processing, and preservation of samples.
- SuperFastPrep-2 uses a unique mechanism mounted on a rotary tool designed to operate at 600 to 4,800 cycles per minute with a fully charged battery. The self-contained device holds two standard 2 mL screw cap Lysing Matrix tubes optimized for a wide selection of sample types. Included with the unit is a rotary handle, two lithium ion rechargeable batteries, charging station, voltage converter, and protective carrying case.
- Applications: Used to homogenize, grind or lyse any samples, the SuperFastPrep-2 portable homogenizer thoroughly and quickly lyses any tissue and cell type, allowing easy and reproducible isolation of stable RNA, active proteins and full-length genomic DNA.
- Sample Volume: Up to 200 mg in 1 mL buffer
- Capacity: 2 x 2 mL Tube
- 90/240 V for lab version, cordless operation for field version.
Recommended Storage
Store at Room Temperature (15–30°C).
Specifications
It can be used to lyse microbes in soil, sediment, sludge, and fecal samples and can effectively process tough-to-lyse fungal, algal, plant, and animal tissues. It can be used at any remote location and in most weather conditions when immediate sample collection, processing, and preservation are required by the researcher. | |
80 to 100 dB | |
Up to 200 mg in 1 mL buffer |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.