Simport™ Scientific SUPERFLO™ Histology Tissue Cassette, One-Piece Molded, Taped
This model is a one-piece molded cassette so no separate lids to manage. It has a front opening and a 45° smooth writing surface. It can be opened and closed up to five times guaranteed, securely relocking to avoid specimen loss. Molded from high-density polymer, it keeps specimens safely submerged in liquid.
$397.61 - $397.61
Specifications
Packaging Type Taped
Quantity 25 Stacks of 40
Pore Style 0.91 x 3.67 mm
Product Type Tissue Cassette
View More Specs
Products 11
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
23045727
View Documents
Simport™ Scientific
M4302T
White
Case of 1000 Each for $397.61
23045728
View Documents
Simport™ Scientific
M4303T
Pink
Case of 1000 Each for $397.61
23045729
View Documents
Simport™ Scientific
M4304T
Green
Case of 1000 Each for $397.61
23045730
View Documents
Simport™ Scientific
M4305T
Yellow
Case of 1000 Each for $397.61
23045731
View Documents
Simport™ Scientific
M4306T
Blue
Case of 1000 Each for $397.61
23045732
View Documents
Simport™ Scientific
M4307T
Peach
Case of 1000 Each for $397.61
23045733
View Documents
Simport™ Scientific
M4308T
Tan
Case of 1000 Each for $397.61
23045734
View Documents
Simport™ Scientific
M4309T
Gray
Case of 1000 Each for $397.61
23045735
View Documents
Simport™ Scientific
M43010T
Lilac
Case of 1000 Each for $397.61
23045736
View Documents
Simport™ Scientific
M43011T
Orange
Case of 1000 Each for $397.61
23045737
View Documents
Simport™ Scientific
M43012T
Aqua
Case of 1000 Each for $397.61
Description
Description
  • Unique lateral vents, fluids can access tissue from all 4 sides
  • Basket style for small tissue management
  • One-piece molded with front opening
  • Smooth 45° angle writing surface
  • Available in 11 colors
  • This cassette model is supplied in taped stacks for use with ink jet printers
  • IVDR
  • CE
Specifications
Specifications
Taped
25 Stacks of 40
0.91 x 3.67 mm
Tissue Cassette