Epredia™ Superfrost™ Plus Adhesion Control Microscope Slide
Histology Superfrost Slide with designated area for known control sample. Available in Standard, or Extended Tab for automated and manual staining.
Supplier:  Epredia™ I8432PLUS001
 View more versions of this product
Catalog No. 22-042-941
Explore available promotions
Description
Description
  • Superfrost Plus adhesion coating
  • 75 x 25 x 1 mm
  • Frosted control box outline for placing control samples
  • Available in Standard, or Extended Tab
Specifications
Specifications
144
Superfrost Plus Adhesion
Standard Superfrost Tab