Superior Glove™ Dexterity™ Nitrile Dipped Work Gloves
Seamless knit cotton shell is comfortable and flexible, with a special additive in the nitrile coating to make gloves 100% biodegradable.
Supplier:  Superior Glove™ S15NT8
 View more versions of this product
Catalog No. 29-635-2036
Explore available promotions
Description
Description
  • Nitrile dipped palm coating offers abrasion and puncture resistance
  • An additive in the coating makes gloves 100% biodegradable
  • Seamles knit cotton shell is comfortable and flexible
  • CFIA compliant
Specifications
Specifications
ANSI 4
Nitrile
Specific
Dip-coated Gloves
Automotive Assembly, Maintenance, and Metal Stamping
ANSI/ISEA 105-16 Level 2
Seamless Knit
Blue, White
Cotton
8
Knit
S15NT