missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Superior Glove™ Dexterity™ Nitrile Dipped Work Gloves
Seamless knit cotton shell is comfortable and flexible, with a special additive in the nitrile coating to make gloves 100% biodegradable.
$50.38 - $59.46
Specifications
ANSI Abrasion Rating | ANSI 4 |
---|---|
Construction | Seamless Knit |
Coating Material | Nitrile |
Color | Blue, White |
Hand | Specific |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
29-635-2035
|
Superior Glove™
S15NT6 |
6 |
Dozen of 12 Each for $54.10
|
|
|||||
28-709-59
|
Superior Glove™
S15NT7 |
7 |
Dozen of 12 Each for $50.86
|
|
|||||
29-635-2036
|
Superior Glove™
S15NT8 |
8 |
Dozen of 12 Each for $51.00
|
|
|||||
28-709-60
|
Superior Glove™
S15NT9 |
9 |
Dozen of 12 Each for $54.43
|
|
|||||
29-635-2037
|
Superior Glove™
S15NT10 |
10 |
Dozen of 12 Each for $50.38
|
|
|||||
29-635-6907
|
Superior Glove™
S15NT11 |
11 |
Dozen of 12 Each for $59.46
Case of 12 Dozen for $649.68
|
|
|||||
Description
- Nitrile dipped palm coating offers abrasion and puncture resistance
- An additive in the coating makes gloves 100% biodegradable
- Seamles knit cotton shell is comfortable and flexible
- CFIA compliant
Specifications
ANSI 4 | |
Nitrile | |
Specific | |
Dip-coated Gloves | |
Knit | |
S15NT |
Seamless Knit | |
Blue, White | |
Cotton | |
Automotive Assembly, Maintenance, and Metal Stamping | |
ANSI/ISEA 105-16 Level 2 |