missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ SuperScript™ IV RT-LAMP Master Mix DFS Item

Invitrogen SuperScript IV RT-LAMP Master Mix is a ready-to-use solution optimized for fast, sensitive, and simple RNA target detection in loop-mediated isothermal amplification (LAMP) reactions.

Supplier:  Invitrogen™ A51802

 View more versions of this product

Catalog No. A51802


Explore available promotions

Description

Description

Invitrogen SuperScript IV RT-LAMP Master Mix is a ready-to-use solution optimized for fast, sensitive, and simple RNA target detection in loop-mediated isothermal amplification (LAMP) reactions. With this convenient master mix formulation, you can perform isothermal amplification from both RNA or DNA targets in a single tube format. 

Benefits of SuperScript IV RT-LAMP Master Mix
• RNA and DNA target detection using Bst DNA polymerase for isothermal amplification in as little as five minutes
• Robust and specific cDNA synthesis using SuperScript IV Reverse Transcriptase
• Enhanced specificity and sensitivity with buffer formulation and RNaseOUT recombinant ribonuclease inhibitor
• Compatible with real-time and endpoint detection—includes SYTO 9 dye for real-time detection
• Flexible design provides several options for evaluating results, including real-time and endpoint detection methods

SuperScript IV RT-LAMP Master Mix includes an evolved Bst DNA polymerase mutant with a fast reaction speed and increased sensitivity and tolerance to inhibitors, as well as SuperScript IV Reverse Transcriptase with increased processivity, thermostability, and inhibitor resistance. To prevent RNA degradation by carry-over RNases, the master mix includes the potent, noncompetitive RNaseOUT Recombinant Ribonuclease Inhibitor. Our proprietary reaction buffer composition is optimized to increase specificity by delaying no-template control (NTC) amplification time.

Specifications

Specifications

Master Mix
Bst DNA Polymerase

• 2X SuperScript IV RT-LAMP Master Mix (4 x 1.25 mL)
• SYTO Green Fluorescent Nucleic Acid Stain (30 μL)

Store at –20°C and protect from light.

65°C
Dry Ice
SYTO 9 Green Fluorescent Nucleic Acid Stain
5 to 15 minutes
LAMP, RT-LAMP
4 x 1.25 mL
400 Reactions
SuperScript IV
Real-time and End-point Detection
2X
SDS
Documents

Documents

Promotions

Promotions