missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ SuperSignal™ ELISA Pico Chemiluminescent Substrate

HRP substrate optimized to generate an intense light signal and provide exception performance in luminometer-based assays.

$640.14 - $1231.43

Specifications

Product Type ELISA Substrate
Detection Method Chemiluminescence
Product Line SuperSignal™
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase)
View More Specs

Products 2
Catalog Number Mfr. No. Kit Contents Quantity Price Quantity  
Catalog Number Mfr. No. Kit Contents Quantity Price Quantity  
PI37069
View Documents
Thermo Scientific™
37069
Stable Peroxide Buffer, 125mL, Luminol/Enhancer, 125mL 250 mL Kit
Each for $1,231.43
 
PI37070
View Documents
Thermo Scientific™
LSG37070
Stable Peroxide Buffer, 50mL, Luminol/Enhancer, 50mL 100 mL Kit
Pack for $640.14
 
Description

Description

Thermo Scientific™ SuperSignal ELISA Pico Chemiluminescent Substrate is optimized to generate an intense light signal and provide exception performance in luminometer-based assays.

SuperSignal ELISA Pico Substrate provides chemiluminescent detection of horseradish peroxidase (HRP) activity for ELISA methods based on HRP-conjugated antibodies and probes. These ELISAs can take place in either a test tube or a microplate and are quantified by measuring relative light units (RLU) in a luminometer.

Highlights:

  • Immediate light generation – intense signal is produced immediately at room temperature or at 37°C

  • High signal-to-noise ratio – minimal background

  • Low picogram sensitivity – detect proteins in your ELISAs down to the picogram levels

  • Room temperature storage – a consistent product with ambient shipping and no need to store at 4°C

  • 8-hour working solution stability – consistent performance of the working solution over an 8-hour period with only a 10% decrease in activity at 24 hours

  • Flexible – signal can be read in black or white opaque plates

  • Emits light at 425nm – measurable by luminometers

Includes:

Luminol/Enhancer Solution (125mL); Stable Peroxide Buffer (125mL)

Specifications

Specifications

ELISA Substrate
SuperSignal™
HRP (Horseradish Peroxidase)
Chemiluminescence
Chemical Substrate
Upon receipt store product at room temperature. Product shipped at ambient temperature.Introduction
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.