missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ SuperSignal™ West Pico Complete Rabbit IgG Detection Kit

Combines SuperSignal West Pico substrate with HRP-conjugated rabbit or mouse secondary antibodies for a convenient, easy-to-use Western blot detection kit.

Supplier:  Thermo Scientific™ 34084

Catalog No. PI34084


Explore available promotions

Description

Description

Thermo Scientific™ SuperSignal Western Blotting Kits are ideal for busy researchers or people new to Western Blotting.

All the components of these Western blotting kits are optimized for compatibility the SuperSignal West Pico Chemiluminescent Substrate and are available for the detection of mouse and rabbit secondary primary antibodies. All SuperSignal Western Blotting kits come with complete, detailed instructions for those new to the technique of Western Blotting and abbreviated protocols for the experienced user. The kits are also available in a standard format (substrate and optimized HRP-conjugates) or a complete format which also includes the Thermo Scientific SuperBlock Buffer and BupH Tris Buffered Saline.

Highlights:

  • Standard and complete kits – choose kits with just secondary antibody and chemiluminescent substrate or kits that also include blocking buffer and wash buffer

  • Mouse and rabbit kits – choose kits with goat anti-mouse IgG or goat anti-rabbit IgG secondary antibody conjugates

  • Instructions – complete, detailed manual with recommended antibody dilutions

Ordering Information:

Each kit provides sufficient reagents to process 50 miniblots or 4000cm2 of membrane.

Includes:

Standard Detection Kits Include: West Pico Substrate (luminol/enhancer solution and stable peroxide solution), HRP-conjugated anti-rabbit IgG, anti-mouse IgG, or biotin-binding protein. Complete Detection Kits also include SuperBlock Blocking Buffer and TBS wash buffer.

Specifications

Specifications

SuperSignal West Pico Complete Rabbit IgG Detection Kit
Chemiluminescence
Nitrocellulose, PVDF
1.5 L
Horseradish Peroxidase
50 mini blots (4000 sq. cm of membrane)
Rabbit IgG Detection Kit
Western Blot
SuperSignal™
HRP
Rabbit
Sufficient For: ∼62 mini blots probed with rabbit primary antibodies
• SuperSignal West Pico Luminol/Enhancer Solution, 250 mL
• SuperSignal West Pico Stable Peroxide Solution, 250 mL
• Peroxidase Conjugated Goat anti-Rabbit IgG (H + L), 2 mL
• SuperBlock Blocking Buffer, 1 L
• TBS Packs (each makes 500 mL), 4 packs
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.