Fisherbrand™ Superslip™ Cover Slips
Developed to minimize sticking, these cover slips are for use in automated coverslippers.
Supplier:  Fisherbrand™ 24X501WYOO811AT
 View more versions of this product
Catalog No. 12541056CA
Explore available promotions
Description
Description
All Superslip™ Cover Slips feature a high-spectral transmission quality and finely adaptive refractive index suited for microscopy.
  • Uniquely developed to reduce sticking
  • Can be used with automated coverslippers or for manual use in humid environments
  • Built-in moisture resistance
  • Manufactured with colorless borosilicate glass
  • Available in a variety of sizes and thicknesses
Specifications
Specifications
0.13 to 0.17 mm (#1)
50 x 24 mm