Supertek Scientific Acrylic Burets
Transparent buret with leakproof stopcock fitted with self lubricating PTFE plug.
Supplier: Supertek Scientific A101181
Description
- Buret is individually calibrated to give maximum accuracy
- Acrylic body buret is unaffected by dilute mineral acids and bases except Hydroflouric Acid Ammonium Hydroxide
Specifications
25 mL | |
Acrylic | |
PTFE | |
Buret |