Supertek Scientific Tirrill Burners
Allows adjustment of both gas and air supply.
Supplier: Supertek Scientific CH106243
Description
- Provides hotter flame
- With brass needle valve to adjust gas supply
- Air supply regulated by adjustable air ports
- Temp range: can reach upto 1550°C
Specifications
Tirrill Bunsen Burner | |
Allows Adjustment of Gas and Air Supply | |
1550°C |
Natural Gas | |
Brass Needle Valve with Adjustable Air Port |