Supertek Scientific Wickless Alcohol Burners
Safe alternative to conventional alcohol burners
Supplier: Supertek Scientific CH10652
Description
Safe alternative to conventional alcohol burners
- Comprising chrome plated tank having screw top with rubber gasket
- Copper torch used instead of wick
- Full leakproof
Specifications
Alcohol Burner | |
100 mL | |
3.25 in. | |
Comprising Chrome-Plated Tank Having screw Top with Rubber Gasket |
0.02 gal. | |
Alternative to Conventional Alcohol Burner | |
8.25 cm |