missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Suplatast Tosylate 98.0+%, TCI America™
$85.75 - $508.89
Chemical Identifiers
| CAS | 94055-76-2 |
|---|---|
| Molecular Formula | C23H33NO7S2 |
| Molecular Weight (g/mol) | 499.64 |
| MDL Number | MFCD00867604 |
| InChI Key | RYVJQEZJUFRANT-UHFFFAOYNA-N |
| Synonym | suplatast tosilate, suplatast tosylate, suplatast, suplatast tosilate inn, ccris 6596, suplatastum tosilas inn-latin, tosilate de suplatast inn-french, tosilato de suplatast inn-spanish, +--2-4-3-ethoxy-2-hydroxypropoxy phenyl carbamoyl ethyldimethylsulfonium p-tosylate |
| PubChem CID | 71773 |
| IUPAC Name | (2-{[4-(3-ethoxy-2-hydroxypropoxy)phenyl]carbamoyl}ethyl)dimethylsulfanium 4-methylbenzene-1-sulfonate |
| SMILES | CC1=CC=C(C=C1)S([O-])(=O)=O.CCOCC(O)COC1=CC=C(NC(=O)CC[S+](C)C)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
S0875100MG
|
TCI America
S0875100MG |
100 mg |
Each for $85.75
|
|
|||||
|
S08751G
|
TCI America
S08751G |
1 g |
Each for $508.89
|
|
|||||
Chemical Identifiers
| 94055-76-2 | |
| 499.64 | |
| RYVJQEZJUFRANT-UHFFFAOYNA-N | |
| 71773 | |
| CC1=CC=C(C=C1)S([O-])(=O)=O.CCOCC(O)COC1=CC=C(NC(=O)CC[S+](C)C)C=C1 |
| C23H33NO7S2 | |
| MFCD00867604 | |
| suplatast tosilate, suplatast tosylate, suplatast, suplatast tosilate inn, ccris 6596, suplatastum tosilas inn-latin, tosilate de suplatast inn-french, tosilato de suplatast inn-spanish, +--2-4-3-ethoxy-2-hydroxypropoxy phenyl carbamoyl ethyldimethylsulfonium p-tosylate | |
| (2-{[4-(3-ethoxy-2-hydroxypropoxy)phenyl]carbamoyl}ethyl)dimethylsulfanium 4-methylbenzene-1-sulfonate |
Specifications
| 94055-76-2 | |
| C23H33NO7S2 | |
| 100 mg | |
| RYVJQEZJUFRANT-UHFFFAOYNA-N | |
| (2-{[4-(3-ethoxy-2-hydroxypropoxy)phenyl]carbamoyl}ethyl)dimethylsulfanium 4-methylbenzene-1-sulfonate | |
| 71773 | |
| ≥98.0% (HPLC,N) | |
| Suplatast Tosylate |
| 87°C | |
| MFCD00867604 | |
| suplatast tosilate, suplatast tosylate, suplatast, suplatast tosilate inn, ccris 6596, suplatastum tosilas inn-latin, tosilate de suplatast inn-french, tosilato de suplatast inn-spanish, +--2-4-3-ethoxy-2-hydroxypropoxy phenyl carbamoyl ethyldimethylsulfonium p-tosylate | |
| CC1=CC=C(C=C1)S([O-])(=O)=O.CCOCC(O)COC1=CC=C(NC(=O)CC[S+](C)C)C=C1 | |
| 499.64 | |
| 499.64 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : WR7906000
TSCA : No