3M™ Supplied-Air Respirator Air-Control Devices
Air-control devices provide positive pressure and regulate the flow and temperature of compressed air.
Supplier: 3M™ 7000126279
Alert:
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Description
- All heating, cooling, and connector assemblies include waist belt and tube with quick-disconnect fitting
- 3M Catalog# V-100 , 3M Catalog# V-300 , 3M Catalog# V-400
Specifications
Connector Assembly | |
Flow Control Valve | |
Blasting, assembly, mechanical, painting, metal repair, cleaning | |
W-3020 Low Pressure Hose, L Series Loose Fitting Headgear, Whitecap Helmets And H Series Hoods, 3M Belt Mounted Supplied Air Respirator |