missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Synthware™ Support Stand

Support Stand features a cast iron base with two holes designed to fit the included chrome plated rod.

$117.38 - $230.38

Specifications

Material Cast Iron, Steel
Diameter (English) Rod 0.40 in.
Diameter (Metric) Rod 10 mm
Product Type Support Stand
Shape Rectangular
View More Specs

Products 3
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
31502257
View Documents
Synthware™
J390260
200 x 125 mm (Base)
Each for $117.38
 
31502258
View Documents
Synthware™
J390270
250 x 160 mm (Base)
Each for $167.91
 
31502259
View Documents
Synthware™
J390280
315 x 200 mm (Base)
Each for $230.38
 
Description

Description

  • Cast iron base with chrome plated rod.
  • Base features two holes designed to fit the included rod.
Specifications

Specifications

Cast Iron, Steel
10 mm
Rectangular
Chrome Plated
0.40 in.
Support Stand
Blue
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'