missing translation for 'onlineSavingsMsg'
Learn More

Suramin hexasodium salt, 98%, Thermo Scientific Chemicals

$348.44 - $348.44

Chemical Identifiers

CAS 129-46-4
Molecular Formula C51H34N6Na6O23S6
Molecular Weight (g/mol) 1429.15
MDL Number MFCD00210217
InChI Key VAPNKLKDKUDFHK-UHFFFAOYSA-H
Synonym suramin sodium, suramin hexasodium, suramin hexasodium salt, suramin sodium salt, antrypol, moranyl, naganinum, naphuride sodium, fourneau 309, suramine sodium salt
PubChem CID 8514
IUPAC Name hexasodium;8-[[4-methyl-3-[[3-[[3-[[2-methyl-5-[(4,6,8-trisulfonatonaphthalen-1-yl)carbamoyl]phenyl]carbamoyl]phenyl]carbamoylamino]benzoyl]amino]benzoyl]amino]naphthalene-1,3,5-trisulfonate
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[Na+].CC1=CC=C(C=C1NC(=O)C1=CC(NC(=O)NC2=CC(=CC=C2)C(=O)NC2=CC(=CC=C2C)C(=O)NC2=C3C(C=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C2)S([O-])(=O)=O)=CC=C1)C(=O)NC1=C2C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C1)S([O-])(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC328540500
SDS View Documents
Thermo Scientific Chemicals
328540500
50 mg Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

129-46-4
1429.15
VAPNKLKDKUDFHK-UHFFFAOYSA-H
8514
[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].CC1=CC=C(C=C1NC(=O)C1=CC(NC(=O)NC2=CC(=CC=C2)C(=O)NC2=CC(=CC=C2C)C(=O)NC2=C3C(C=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C2)S([O-])(=O)=O)=CC=C1)C(=O)NC1=C2C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C1)S([O-])(=O)=O
C51H34N6Na6O23S6
MFCD00210217
suramin sodium, suramin hexasodium, suramin hexasodium salt, suramin sodium salt, antrypol, moranyl, naganinum, naphuride sodium, fourneau 309, suramine sodium salt
hexasodium;8-[[4-methyl-3-[[3-[[3-[[2-methyl-5-[(4,6,8-trisulfonatonaphthalen-1-yl)carbamoyl]phenyl]carbamoyl]phenyl]carbamoylamino]benzoyl]amino]benzoyl]amino]naphthalene-1,3,5-trisulfonate

Specifications

129-46-4
97.5% min.
97.5% min. (HPLC)
C51H34N6Na6O23S6
50 mg
suramin sodium, suramin hexasodium, suramin hexasodium salt, suramin sodium salt, antrypol, moranyl, naganinum, naphuride sodium, fourneau 309, suramine sodium salt
VAPNKLKDKUDFHK-UHFFFAOYSA-H
hexasodium;8-[[4-methyl-3-[[3-[[3-[[2-methyl-5-[(4,6,8-trisulfonatonaphthalen-1-yl)carbamoyl]phenyl]carbamoyl]phenyl]carbamoylamino]benzoyl]amino]benzoyl]amino]naphthalene-1,3,5-trisulfonate
8514
98%
Suramin hexasodium salt
White
Authentic
Glass bottle
MFCD00210217
15, 9135
Solubility in water: soluble in water
[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].CC1=CC=C(C=C1NC(=O)C1=CC(NC(=O)NC2=CC(=CC=C2)C(=O)NC2=CC(=CC=C2C)C(=O)NC2=C3C(C=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C2)S([O-])(=O)=O)=CC=C1)C(=O)NC1=C2C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C1)S([O-])(=O)=O
1429.15
1429.1
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-949-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only