missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ SureOne™ Beveled Pipette Tips, Universal Fit, Without Graduations

Offers a full line of universal fit beveled pipet tips

$63.61 - $63.61

Specifications

Volume (Metric) 1 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Autoclavable Autoclavable
Length (Metric) 51 mm
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Tip Style Color Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Tip Style Color Price Quantity  
02-707-450 Edge
View Documents Promotion Details
N/A 1 to 200 μL Beveled Clear
Pack of 1000 Each for $63.61
Case of 10 Pack for $636.10
Edge
 
02-707-451 Edge
View Documents Promotion Details
N/A 1 to 200 μL Beveled Yellow
Pack of 1000 Each for $63.61
Case of 10 Pack for $636.10
Edge
 
Description

Description

Offers a full line of universal fit beveled pipet tips

  • All racked sterile tips are shrink wrapped and certified free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin, and DNA
  • All nonsterile racked and bulk nonfiltered tips are certified free of RNase, DNase, and DNA

SureOne Rack Features

  • Flush Fit Inserts sit flush with rack, allowing easy multichannel pipetting
  • Notched clasp props lid open for autoclave venting
  • Sure Stack lid rim prevents racks from sliding around when stacked and carted around a lab
  • Safe-Lock latch allows for one-handed open and closing ability and won't pop open when dropped
  • Sure-snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Reversible flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
  • Recycle logo molded into all rack components
SureOne Insert Features
  • Inserts are easy to place onto and remove from rack; clip onto rack for a secure fit
  • Lot traceable — all inserts contain the Fisherbrand part number, tip volume, lot number
  • Indented insert holes inhibit tip movement, stabilizing them for multichannel work
  • Contact clip prevents unnecessary contact with the tips while clipping in inserts
  • Multichannel design; all inserts are 96-well format
  • Robust non-pliable design provides secure seals on every tip
SureOne Reload Packaging Features
  • Product information displayed on box
  • Dust flap prevents debris from contaminating the product
  • Finger knockouts allow easy access to slide out product
  • Canyon cut package allows for easy tip insert removal
  • Reload packaging design saves 58% bench space vs. racked packaging; reduces plastic waste by 52%

Specifications

Specifications

1 to 200 μL
Non-sterile
51 mm
Bulk
Universal Fit
SureOne
Non-filtered
Autoclavable
DNase, RNase, and DNA Free
Beveled
No
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'