Fisherbrand™ SureStack Pipet Tips
Pipetting station with high quality tips.
 View more versions of this product
Catalog No. 02-100-502
Explore available promotions
Description
Description
  • Ideal for use with multi-channel pipetters, both 8 and 12 channel
  • Decks snap together and will not lift during pipetting
  • Recessed wells keep tips stable for mounting
  • Saves bench space
  • Lid can be adjusted to expose only 96 tips
  • Wide base design prevents stack from tipping
  • Trays manufactured from recycled plastic
  • Autoclaving not recommended: available presterilized
  • 200μL pipet tip features a soft fit collar, graduation marks at 10, 50 and 100μL
Specifications
Specifications
200 μL
Natural
Standard Length
Yes
Recycled Plastic
Non-sterile
Not Autoclavable
Universal Fit
SureStack