Surgical Design Graduated Scalpel Handles
Offers etched graduated markings for convenient measurements
Supplier:  Surgical Design 270G
 View more versions of this product
Catalog No. 22-079-686
Explore available promotions
Description
Description
  • Designed with metric scale on handle
  • Measurements can be made in centimeters and millimeters
Specifications
Specifications
# 3