Surgical Design Graduated Scalpel Handles
Offers etched graduated markings for convenient measurements
Supplier: Surgical Design 271G
Description
- Designed with metric scale on handle
- Measurements can be made in centimeters and millimeters
Specifications
# 4 |