Learn More
Surgical Design Surgical Knife Set; Set of 8 Scalpel Handles
Surgical Design™ Surgical Knife Set includes the most common and widely used scalpel handles in the field.
Supplier:  Surgical Design SSET8
Catalog No. 22-079-771
Only null left
Explore available promotions
Description
Description
  • Premium grade scalpel handles. Set includes
  • (1) Scalpel handle, size #3
  • (1) Scalpel handle, size #4
  • (1) Scalpel handle, size #7
  • (1) Scalpel handle, size #3, Extra-long handle
  • (1) Scalpel handle, size #4, Extra-long handle
  • (1) Scalpel handle with measuring scales, size #3
  • (1) Scalpel handle with measuring scales, size #4
  • (1) Blade remover
Specifications
Specifications
Stainless Steel
Varies between Handles
Satin, Anti-glare
Autoclavable
Surgical Knife Handle Set
Compatible with 10 and 20 Series Scalpel Blade
Reusable
Yes
Product Suggestions
Product Suggestions