Learn More
MilliporeSigma™ Supelco™ Swagelok™ Nut, Front and Back Ferrule Set, 400-S, Stainless Steel, 1/4 in.
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic.
Supplier:  MilliporeSigma™ Supelco™ SGMA22029
Catalog No. 111005035
Only null left
Explore available promotions
Description
Description
The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. Brass ferrules are used with copper tubing whereas stainless-steel ferrules are used with stainless-steel tubing.
Specifications
Specifications
5/Pk.
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Swagelok is a registered trademark of Swagelok Company