Learn More
Agilent BioTek Synergy Neo2 Hybrid Multi-Mode Microplate Reader
BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. For Research Use Only. Not for use in diagnostic procedures.
$61652.81 - $203530.15
Specifications
Absorbance Range | 230 to 999 nm, 1 nm increments |
---|---|
Accuracy | Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
CO2 Concentration Range | 0 to 20% |
Depth (Metric) | 52.5 cm |
Catalog Number | Mfr. No. | Model | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Includes | Price | Quantity | ||||
BTNEO2
|
Agilent BioTek
N2SN |
NEO2 | Monochromator: ABS; Filter: Dual filter top FI/FP/TRF and LUM |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTNEO2M
|
Agilent BioTek
N2MSN |
NEO2M | Monochromator: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTNEO2MB
|
Agilent BioTek
N2MBSN |
NEO2MB | Monochromater: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTNEO2S
|
Agilent BioTek
N2SSN |
NEO2S | Monochromater: ABS; Filter: Single filter top FI/FP/TRF and LUM |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTNEO2SM
|
Agilent BioTek
N2SMSN |
NEO2SM | Monochromater: ABS, FI and LUM; Filter: Single filter top FI/FP/TRF and LUM |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTNEO2SMB
|
Agilent BioTek
N2SMBSN |
NEO2SMB | Monochromater: ABS, FI and LUM; Filter: Single filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTNEO2MBT
|
Agilent BioTek
N2MBTSN |
NEO2MBT | Monochromater: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF, TRF Laser |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTNEO2MON
|
Agilent BioTek
N2MONSN |
NEO2MON | Monochromater: ABS, FI and LUM |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTNEO2MT
|
Agilent BioTek
N2MTSN |
NEO2MT | Monochromator: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, TRF Laser |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTNEO2T
|
Agilent BioTek
N2TSN |
NEO2T | Monochromater: ABS; Filter: Dual filter top FI/FP/TRF and LUM, TRF Laser |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
NEO2ALPHABT
|
Agilent BioTek
N2ABTSN |
NEO2ALPHABT | Monochromater: ABS; Filter: Dual filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF, Alpha Laser, TRF Laser |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
NEO2ALPHAT
|
Agilent BioTek
N2ATSN |
NEO2ALPHAT | Monochromater: ABS; Filter: Dual filter top FI/FP/TRF and LUM, Alpha Laser, TRF Laser |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
NEO2MALPHAT
|
Agilent BioTek
N2MATSN |
NEO2MALPHAT | Monochromater: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, Alpha Laser, TRF Laser |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
NE2MALPHABT
|
Agilent BioTek
N2MABTSN |
NEO2MALPHABT | Monochromater: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF, Alpha Laser, TRF Laser |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
NE2SMALPHAB
|
Agilent BioTek
N2SMABSN |
NEO2SMALPHAB | Monochromater: ABS, FI and LUM; Filter: Single filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF, Alpha Laser |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
NEO2MALPHA
|
Agilent BioTek
N2MASN |
NEO2MALPHA | Monochromater: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, Alpha Laser |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
NEO2MALPHAB
|
Agilent BioTek
N2MABSN |
NEO2MALPHAB | Monochromater: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF, Alpha Laser |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
NEO2SALPHA
|
Agilent BioTek
N2SASN |
NEO2SALPHA | Monochromater: ABS; Filter: Single filter top FI/FP/TRF and LUM, Alpha Laser |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
NEO2SMALPHA
|
Agilent BioTek
N2SMASN |
NEO2SMALPHA | Monochromater: ABS, FI and LUM; Filter: Single filter top FI/FP/TRF and LUM, Alpha Laser |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. Variable bandwidth quad monochromators, sensitive high transmission filter-based optics, laser TRF, and up to 4 PMTs provide ultrafast measurements.
Advanced environmental controls, including CO2/O2 control, incubation to 70°C, and variable shaking support live cell assays. BioStack Neo delivers walk-away automation, high throughput, barcode-labeled filter cubes to streamline workflows and limit errors. Powerful Gen5 software provides complete reader control, powerful data analysis, and LIMS integration.
Specifications
230 to 999 nm, 1 nm increments | |
CE and TUV marked. RoHS compliant. | |
52.5 cm | |
±0.2°C at 37°C | |
0 to 4 OD | |
HTS, biomarker, nucleic acid, protein quantification, rapid kinetics, HTRF, SNP, TR FRET, BRET, Cytokines, ELISA, FP | |
41 cm | |
250 W max. | |
78 lb. | |
15.4 in. | |
Photodiode, Fluorescence/Alpha: Dual PMT or Single PMT (option) | |
Xenon Flash |
Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD | |
0 to 20% | |
20.7 in. | |
Linear, Orbital, Double Orbital | |
5 to 1000 μL, 1 μL increments | |
16.1 in. | |
Multi-Mode Microplate Reader | |
0.0001 OD | |
35 kg | |
39 cm | |
Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option) |