missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Synth-a-Freeze™ Cryopreservation Medium DFS Item

Synth-a-Freeze™ Cryopreservation Medium

Supplier:  Gibco™ A1254201

Catalog No. A1254201

Explore available promotions

Description

Description

Synth-a-Freeze™ is a defined, liquid cryopreservation medium containing 10% dimethylsulfoxide (DMSO) without antibiotics, antimycotics, hormones, growthfactors, serum or protein and buffered with HEPES and sodium bicarbonate. Synth-a-Freeze is intended for freezing and storing a variety of mammalian cell types including human keratinocytes, embryonic stem cells, neural stem cells and mesenchymal stem cells. It should not be used for melanocytes.
Specifications

Specifications

Store in freezer (-5 to -20°C).
Animal Origin-free
Cryopreservation Media
Sterile
< 1 EU/ml
Qualified for Further Manufacturing
Dry Ice
Keratinocyte Cell, Embryonic Stem Cell, Neural Stem Cell, Mesenchymal Stem Cell
Liquid
Serum-free
HEPES, Phenol Red, DMSO (10%)
Gibco™, Synth-a-Freeze™
50 mL
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.