Synthware™ Addition Funnels with Pressure Equalizing Arm and 1:5 PTFE Stopcock
Cylindrical style addition funnel with pressure equalizing arm and PTFE stopcock
Supplier:  Synthware™ F621425
 View more versions of this product
Catalog No. 31501693
Explore available promotions
Description
Description
  • Top outer and lower inner standard taper joints of same size
Specifications
Specifications
25 mL
14/20
Pressure-equalizing Funnel