Synthware™ Addition Funnels with Pressure Equalizing Arm and 1:5 PTFE Stopcock
Cylindrical style addition funnel with pressure equalizing arm and PTFE stopcock
Supplier: Synthware™ F62241L
Description
- Top outer and lower inner standard taper joints of same size
Specifications
1 L | |
24/40 | |
Pressure-equalizing Funnel |