Synthware™ Allihn Condenser
With joints at the top and the bottom
Supplier:  Synthware™ C254300
 View more versions of this product
Catalog No. 31500793
Explore available promotions
Description
Description
  • With standard taper outer joint at top and lower inner drip tip joint
  • Both joints of same size
Specifications
Specifications
Hose connection: 8mm O.D.
None
Allihn Condenser
Borosilicate Glass (3.3)
24/40
Distillation
300 mm
Distillation Head