Synthware™ Allihn Condenser for Soxhlet Extractor
For use with Soxhlet extraction apparatus
Supplier:  Synthware™ C243445
 View more versions of this product
Catalog No. 31501073
Explore available promotions
Description
Description
  • Allihn reflux condenser with water cooled inner drip tip joint at bottom
Specifications
Specifications
34/45
Distillation
Allihn Condenser
Glass
None
275mm
190 mm
Soxhlet Extractor