missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Synthware™ Buchner Funnels with Perforated Glass Plate
With sealed in perforated glass disc to support filter media
Supplier: Synthware™ F808350
Description
- Design solves contamination problem with fritted disc and blindness problem with ceramic Buchner funnel
- Note: Not recommended for filtration where pressure would exceed 15 psi
Spécifications
350 mL | |
Clear | |
Buchner Funnel with Perforated Glass Plate |
Borosilicate Glass (3.3) | |
80mm |