Synthware™ Chromatography Columns with Standard Taper Joint, PTFE Stopcock and Fritted Disc
Comes in variety of formulation to provide maximum flexibility, control, convenience, and improved yield
Supplier:  Synthware™ C184322C
 View more versions of this product
Catalog No. 31500869
Explore available promotions
Description
Description
  • WIth standard taper top outer joint
  • Heavy wall design for increased safety
Specifications
Specifications
26 mm
Chromatography Column
254 mm
10 in.
2 mm bore
32 mm