Synthware™ Chromatography Columns with Standard Taper Joint, PTFE Stopcock and Fritted Disc
Comes in variety of formulation to provide maximum flexibility, control, convenience, and improved yield
Supplier: Synthware™ C184461C
Description
- WIth standard taper top outer joint
- Heavy wall design for increased safety
Specifications
40 mm | |
Chromatography Column | |
203 mm |
8 in. | |
2 mm bore | |
46 mm |