Synthware™ Chromatography Columns with Standard Taper Joint and PTFE Stopcock
General purpose flash chromatography columns
$178.79 - $436.91
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) Inner | Diameter (Metric) Outer | Length (English) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) Inner | Diameter (Metric) Outer | Length (English) | Price | Quantity | |||
31500841
|
Synthware™
C184171 |
203 mm | 13.4 mm | 17 mm | 8 in. |
Each for $178.79
|
|
|||
31500842
|
Synthware™
C184172 |
254 mm | 13.4 mm | 17 mm | 10 in. |
Each for $189.00
|
|
|||
31500843
|
Synthware™
C184173 |
305 mm | 13.4 mm | 17 mm | 12 in. |
Each for $199.21
|
|
|||
31500844
|
Synthware™
C184261 |
203 mm | 20 mm | 26 mm | 8 in. |
Each for $178.79
|
|
|||
31500845
|
Synthware™
C184262 |
254 mm | 20 mm | 26 mm | 10 in. |
Each for $189.00
|
|
|||
31500846
|
Synthware™
C184263 |
305 mm | 20 mm | 26 mm | 12 in. |
Each for $199.21
|
|
|||
31500847
|
Synthware™
C184264 |
457 mm | 20 mm | 26 mm | 18 in. |
Each for $239.35
|
|
|||
31500848
|
Synthware™
C184321 |
203 mm | 26 mm | 32 mm | 8 in. |
Each for $221.85
|
|
|||
31500849
|
Synthware™
C184322 |
254 mm | 26 mm | 32 mm | 10 in. |
Each for $239.35
|
|
|||
31500850
|
Synthware™
C184323 |
305 mm | 26 mm | 32 mm | 12 in. |
Each for $257.00
|
|
|||
31500851
|
Synthware™
C184324 |
457 mm | 26 mm | 32 mm | 18 in. |
Each for $307.35
|
|
|||
31500852
|
Synthware™
C184461 |
203 mm | 40 mm | 46 mm | 8 in. |
Each for $222.56
|
|
|||
31500853
|
Synthware™
C184462 |
254 mm | 40 mm | 46 mm | 10 in. |
Each for $239.15
|
|
|||
31500854
|
Synthware™
C184463 |
305 mm | 40 mm | 46 mm | 12 in. |
Each for $257.00
|
|
|||
31500855
|
Synthware™
C184464 |
457 mm | 40 mm | 46 mm | 18 in. |
Each for $307.35
|
|
|||
31500856
|
Synthware™
C184603 |
305 mm | 53 mm | 60 mm | 12 in. |
Each for $333.62
|
|
|||
31500857
|
Synthware™
C184604 |
457 mm | 53 mm | 60 mm | 18 in. |
Each for $370.38
|
|
|||
31500858
|
Synthware™
C184605 |
610 mm | 53 mm | 60 mm | 24 in. |
Each for $436.91
|
|
|||
Description
- WIth standard taper top outer joint
- Heavy wall design for increased safety
Specifications
Chromatography Column | |
2 mm bore |