Synthware™ Concentrator Tube, Graduated
For use with Kuderna-Danish evaporative set-ups
$77.82 - $92.26
Specifications
Product Type Graduated Concentrator Tube
For Use With (Equipment) Kuderna-Danish Evaporation Set-up
For Use With (Application) Organic Chemistry
Standard Taper Joint 19/22
View More Specs
Products 2
Catalog Number Mfr. No. Description Height (Metric) Price Quantity  
Catalog Number Mfr. No. Description Height (Metric) Price Quantity  
31501088
View Documents
Synthware™
C291910
Subdivisions: 0 to 1mL in 0.1; 2 to 10mL in 1.0 125 mm
Each for $77.82
31501089
View Documents
Synthware™
C291925
Subdivisions: 0 to 1mL in 0.1; 2 to 25mL in 1.0 170 mm
Each for $92.26
Description
Description
  • Tube can be stoppered for storage
Specifications
Specifications
Graduated Concentrator Tube
Kuderna-Danish Evaporation Set-up
Organic Chemistry
19/22
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'