Synthware™ Distillation Adapter, 75°
Distillation adapter with standard taper inner joints on each end at a 75 deg. angle
$49.94 - $60.44
Specifications
For Use With (Application) | Organic Synthesis |
---|---|
Material | Borosilicate Glass (3.3) |
Product Type | Glass Joint Adapter |
Quantity | 1 |
Description
Synthware distillation adapter bent at 75 degrees. Both ends are standard taper inner joints and are the same sizeSpecifications
Organic Synthesis | |
Borosilicate Glass (3.3) | |
Glass Joint Adapter | |
1 |