Synthware™ Distillation Condenser with Removable Hose Connections
With indentations at the bottom for supporting packing material
Supplier:  Synthware™ C571120
 View more versions of this product
Catalog No. 31500729
Explore available promotions
Description
Description
  • Jacket permits column to be used as condenser
  • With top outer and lower inner standard taper joints
Specifications
Specifications
180mm
Distillation Condenser
14/20